Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553180 Supplier Invitrogen™ Supplier No. PA553180
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKEFYLSNSE KERYEKEFSQ ERQQEILRRA ARALPIYTTS ASKTIRYCEK CQLIKPDRAH HCSACDSCIL KMDHHCPWVN NCV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.

Catalyzes palmitoylation of Cys residues on target proteins (PubMed:27153536, PubMed:29326245). Catalyzes palmitoylation of Cys residues in the cytoplasmic C-terminus of EGFR, and modulates the duration of EGFR signaling by modulating palmitoylation-dependent EGFR internalization and degradation (PubMed:27153536). Has a preference for acyl-CoA with C16 fatty acid chains (PubMed:29326245). Can also utilize acyl-CoA with C14 and C18 fatty acid chains (PubMed:29326245). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC20
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC20
Gene Accession No. Q5W0Z9
Gene Alias 4933421L13Rik; 5033406L14Rik; AI448102; B230110O18Rik; DHHC-20; DHHC-containing protein 20; HK11; membrane-associated DHHC24 zinc finger protein; Palmitoyltransferase ZDHHC20; probable palmitoyltransferase ZDHHC20; RGD1305755; ZDHHC20; Zinc finger DHHC domain-containing protein 20; zinc finger DHHC-type containing 20; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 20
Gene Symbols ZDHHC20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZDHHC20. Recombinant protein control fragment (Product #RP-91187).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253832
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.