Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567229 Supplier Thermo Scientific Supplier No. PA567229
Only null left
Add to Cart
Add to Cart

ZDHHC22 Polyclonal Antibody

ZDHHC22 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC22
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZDHHC22
Gene Accession No. Q8N966
Gene Alias C14orf59; DHHC-22; Gm262; LOW QUALITY PROTEIN: palmitoyltransferase ZDHHC22; membrane-associated DHHC22 zinc finger protein; palmitoyltransferase ZDHHC22; putative palmitoyltransferase ZDHHC22; ZDHHC22; Zinc finger DHHC domain-containing protein 22; zinc finger DHHC-type containing 22; zinc finger DHHC-type palmitoyltransferase 22; zinc finger, DHHC domain containing 22; zinc finger, DHHC-type containing 22
Gene Symbols ZDHHC22
Host Species Rabbit
Immunogen Recombinant Human ZDHHC22. Recombinant protein control fragment (Product #RP-107866).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283576
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.