Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116794 Supplier Invitrogen™ Supplier No. PA5116794
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Rat lung Immunogen sequence: VNLFFFTLTC GTNPGIITKA NELLFLHVYE FDEVMFPKNV RCSTCDLRKP ARSKHCSVCN WCVHRFDHHC VWVNNCIGAW NIRYFLIYVL TLTASAATVA I.

ZDHHC4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZDHHC4
Gene Accession No. Q5FVR1, Q9D6H5, Q9NPG8
Gene Alias 1810021D01Rik; 2900029I10Rik; DC1; DHHC-4; membrane-associated DHHC4 zinc finger protein; probable palmitoyltransferase ZDHHC4; UNQ5787/PRO19576; Zdhhc4; Zinc finger DHHC domain-containing protein 4; zinc finger DHHC-type containing 4; zinc finger protein 374; zinc finger, DHHC domain containing 4; zinc finger, DHHC-type containing 4; ZNF374
Gene Symbols ZDHHC4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 110-210 of human ZDHHC4 (NP_0605761).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304291, 55146, 72881
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.