Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Zea Mays Stem, Combination Longitudinal Section and Cross Section
SCI_ED

Catalog No. S30173 Shop All Eisco Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
S30173 Cross and Longitudinal Section
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30173 Supplier Eisco™ Supplier No. BS18062
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prepared microscope slide of corn stem. Combination slide of cross section and longitudinal section showing characteristic monocot stem features from two different points of view. Ideal choice for biology classrooms or home scientists.

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic monocot stem features from two different points of view

Specifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Mounting Type Cross and Longitudinal Section
Type Microbiology slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Plant
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.