Learn More
Medchemexpress LLC Zearalenone | 17924-92-4 | MFCD00133085 | 98.9% | 318.36 | C18H22O5 | 100 MG
Supplier: Medchemexpress LLC HY103447100MG
Zearalenone is a Fusarium-derived mycotoxin with estrogenic activity used as a research standard and analytical reference for toxicology, food safety, and assay development.
- High purity 98.85% suitable for analytical applications.
- Molecular formula C18H22O5 and molecular weight 318.36.
- Available as solid (including 100 MG) and as a 10 mM solution in DMSO.
- Appropriate for assay development, method validation, and toxicology studies.
- Stable under recommended storage and handling conditions when used as a reference standard.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.