Learn More
Medchemexpress LLC Zearalenone-13C18 | 911392-43-3 | 98.7% | 336.23 g/mol | 13C18H22O5 | 30 UG
SDP
Supplier:  Medchemexpress LLC HY103447S130UG
Encompass_Preferred
Zearalenone-13C18 is a fully 13C-labeled isotopic internal standard of the mycotoxin zearalenone, intended for use as an analytical reference in GC- and LC-MS quantification. The compound has CAS number 911392-43-3, molecular formula 13C18H22O5, and molecular weight 336.23 g/mol. It is supplied as a pre-weighed quantity for analytical laboratory use.
  • Fully 13C-labeled internal standard for mass spectrometry.
  • High purity (98.7%) to support accurate quantification.
  • Suitable for GC- and LC-MS analytical methods.
  • Supplied as a 30 μg pack for trace-level analyses.
Catalog No. 50-002-46362
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.