Learn More
Medchemexpress LLC Zearalenone | 17924-92-4 | MFCD00133085 | 98.9% | 318.36 | C18H22O5 | 50 MG
Supplier: Medchemexpress LLC HY10344750MG
Zearalenone is a resorcylic acid lactone mycotoxin produced by Fusarium species, supplied as an analytical standard for research into estrogenic activity and mycotoxin contamination. The material is provided as a measured solid and is accompanied by supporting documentation for research and analytical use.
- High purity (98.85%) suitable for analytical applications.
- Molecular formula C18H22O5 and molecular weight 318.36.
- Provided as a 50 mg solid; other pack sizes and a 10 mM DMSO solution are available.
- Includes certificate of analysis and safety data sheet for traceability.
- Intended for research and analytical testing of mycotoxin contamination and estrogenic effects.