Learn More
Medchemexpress LLC Zearalenone (standard) | 17924-92-4 | 98.9% | 318.36 | C18H22O5 | 10 MG

Supplier: Medchemexpress LLC HY103447R10MG
Zearalenone (Standard) is an analytical reference standard of the mycotoxin zearalenone for research and analytical applications, including method development, calibration, and quantification in food, feed, and biological matrices.
- Analytical reference standard for zearalenone
- CAS number 17924-92-4
- Molecular formula C18H22O5; molecular weight 318.36
- Purity 98.9% (reported)
- Supplied as a 10 mg standard for analytical workflows
- Storage: in solvent -80°C up to 6 months, -20°C up to 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.