Learn More
Medchemexpress LLC Zearalenone | 17924-92-4 | MFCD00133085 | 98.9% | 318.4 g/mol | C18H22O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY103447R5MG
Encompass_Preferred
Analytical reference standard of the mycotoxin zearalenone intended for research and analytical applications. Supplied as a 5 mg standard with high purity for method development, calibration, and quality control.
  • Analytical reference standard for mycotoxin testing.
  • Suitable for method development and calibration.
  • High purity (98.9%).
  • White microcrystals or powder form for ease of handling.
Catalog No. 50-002-46912
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.