Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zearalenone, Tocris Bioscience™

Catalog No. 397550
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-755-0 50 mg
39-751-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-755-0 Supplier R&D Systems Supplier No. 3975/50
This item is not returnable. View return policy

Estrogen receptor ligand; mycotoxin

Mycotoxin produced by several species of Fusarium fungi. Binds to the estrogen receptor (ER) (IC50 values are 166 and 240 nM for ERβ and ERα respectively) and stimulates the transcriptional activity of both subtypes at concentrations between 1 - 10 nM. Also stimulates growth of T47D breast cancer cells.

Chemical Identifiers

CAS 17924-92-4
Molecular Formula C18H22O5
Molecular Weight (g/mol) 318.369
InChI Key MBMQEIFVQACCCH-QBODLPLBSA-N
Synonym zearalenone, --zearalenone, trans-zearalenone, f-2 toxin, mycotoxin f2, s---zearalenone, zenone, s-zearalenone, toxin f2, 10s-zearalenone
PubChem CID 5281576
ChEBI CHEBI:10106
IUPAC Name (2E,11S)-15,17-dihydroxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,15,17-tetraene-7,13-dione
SMILES CC1CCCC(=O)CCCC=CC2=CC(=CC(=C2C(=O)O1)O)O

Specifications

Quantity 50 mg
Formula Weight 318.36
Percent Purity >98%
Chemical Name or Material Zearalenone
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.