Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZEB1 Mouse anti-Human, Clone: 1H1F1, Proteintech
SDP

Catalog No. 501736711 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6711 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6711 Supplier Proteintech Group Inc Supplier No. 662791IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

THOC3 is part of the TREX (transcription/export) complex, which includes THO2, HPR1, ALY, and UAP56.TEX1 is part of the TREX (transcription/export) complex, which includes THO2 (MIM 300395), HPR1 (MIM 606930), ALY (MIM 604171), and UAP56 (MIM 606390).

Specifications

Antigen ZEB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1H1F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZEB1
Gene Accession No. P37275
Gene Alias AREB6, BZP, DELTA EF1, delta EF1, Negative regulator of IL2, NIL 2 A, NIL 2 A zinc finger protein, NIL 2A, NIL2A, TCF 8, TCF8, Transcription factor 8, ZEB, ZEB1, ZFHEP, ZFHX1A
Gene Symbols ZEB1
Host Species Mouse
Immunogen ZEB1 Fusion Protein Ag17118
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6935
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.