Learn More
Kingfisher Biotech Inc Zebra Finch M-CSF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1747ZN500
Encompass_Preferred
The Zebra Finch M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Zebra Finch M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Zebra Finch M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Zebra Finch M-CSF Specifications: (Molecular Weight: 23.1 kDa) (Amino Acid Sequence: EQNSYCHQII TERHLAELEE LADTQMQHPG RVSFKFIDKM QLNDSICYVK AAFPLMGKIL ERTEFKENSS NARKMQMVRR MYNSIDENVD PCIRDEDDEE RMLSQMCFKE FTTSPYEMLV LVKDFFQDIK RLLQNQETFE KDCSRVYRRV CPGPRKAGSS PGVGTDPDCN CLSPALPPAT QPSLSAATGR DTAPASTRLP SSL (203)) (Gene ID: 100499187). For research use only. Made in the USA
Catalog No. 50-253-2833
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.