Learn More
Kingfisher Biotech Inc Zebrafish CCL2 (MCP-1) Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1575Z005
Encompass
The Zebrafish CCL2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Zebrafish CCL2 applications are for cell culture, ELISA standard, and Western Blot Control. Zebrafish CCL2 yeast-derived recombinant protein can be purchased in multiple sizes. Zebrafish CCL2 Specifications: (Molecular Weight: 14.3 kDa) (Amino Acid Sequence: PIPINSAKSV CCEATTHRNI PLKQIMSYQW TTSTCPIKAI VFKTIAGREI CVDPQNTLVK KQVAKLDKRR STTALSPKST STTAETPAAT SGTTSSSEST SAKSSPASTS ATALSHEYTS ASTVTFTTAE SHSTSV (136)) (Gene ID: 100538158). For research use only. Made in the USA
Catalog No. 50-253-3143
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.