Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zein from Corn, TCI America™
SDP

Catalog No. Z0001500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0001500G 500 g
Z000125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z0001500G Supplier TCI America Supplier No. Z0001500G

Chemical Identifiers

CAS 9010-66-6
MDL Number MFCD00062414

Specifications

Quantity 500 g
Physical Form Crystalline Powder
Chemical Name or Material Zein from Corn
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.