Learn More
Protective Industrial Products Zenon Ultra-Lyte™ Rimless Safety Glasses, Anti-Scratch / Fogless™ 3Sixty™ Coating
GREENER_CHOICE
Catalog No. 17400112 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-400-112 Gray
17-400-111 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-400-112 Supplier Protective Industrial Products Supplier No. 250140521
  • $107.00
    $64.65 / Dozen of 12
    Save $42.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Rimless Safety Glasses featuring squared lens shape with a slight taper that provides modern styling for the wearer.
  • Squared lens shape with a slight taper, provides modern styling for the wearer
  • Ultra-lightweight design conforms to the head promoting all-day wear
  • Spring fit temples offers a comfortable fit that conforms to the head
  • FogLess™ 3Sixty™ enhanced hydrophobic action resists lens fogging and provides clear vision in extreme working conditions
  • Molded nose bridge design to fit the vast majority of users
  • 10 Base curve lens
  • Meets ANSI Z87.1-2020 standards
Specifications
Lens Coating Anti-scratch, Anti-fog
Color Gray
Lens Tint None
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None
Frame Type Rimless