Learn More
PIP™ Zenon™ Z12 Foam
Catalog No. 19148037 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-148-037 Gray
19-148-036 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-148-037 Supplier PIP™ Supplier No. 25001F021
Add to Cart
Add to Cart
Rimless Safety Glasses feature foam padding seal and anti-scratch / anti-fog coating.
  • Lightweight, rimless design provides all day user comfort
  • Foam padding design forms a seal to keep out dust and airborne particles
  • Thin, flexible temples provide a secure fit without uncomfortable pressure points
  • Molded nose bridge design to fit the vast majority of users
  • 9.5 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Lens Coating Anti-fog, Scratch-resistant
Shape Wraparound Lens
Height (Metric) 50 mm
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) General manufacturing and anywhere eye protection is needed
Frame Color Black
Temple Style Thin
Material Polycarbonate, Nylon, Synthetic Foam
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 99.99% UVA and UVB Light Absorption
Frame Type Rimless
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.