Learn More
PIP™ Zenon Z12R Bi-Focals
Catalog No. 19148024 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Frame Material:
Nylon
Polycarbonate
Magnification Power:
+1.00
+1.25
+1.50
+1.75
+2.00
+2.25
+2.50
+2.75
+3.00
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Material Magnification Power
19-148-024 Polycarbonate +1.75
19-148-021 Polycarbonate +1.00
19-148-014 Nylon +1.25
19-148-022 Polycarbonate +1.25
19-148-015 Nylon +1.50
19-148-023 Polycarbonate +1.50
19-148-016 Nylon +1.75
19-148-017 Nylon +2.00
19-148-025 Polycarbonate +2.00
19-148-018 Nylon +2.25
19-148-026 Polycarbonate +2.25
19-148-019 Nylon +2.50
19-148-027 Polycarbonate +2.50
19-148-028 Polycarbonate +2.75
19-148-029 Polycarbonate +3.00
19-148-013 Nylon +1.00
19-148-020 Nylon +3.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 19-148-024 Supplier PIP™ Supplier No. 250270017
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
PIP™ Zenon Z12R Bi-Focals feature rubber tips on the temples to provide comfort behind the ear, as well as a secure fit.
  • Lightweight, rimless design for user comfort
  • Flexible, rubber tip temples provide a secure fit without uncomfortable pressure points
  • Variety of lenses to fit applications
  • Molded nose bridge design to fit the vast majority of users
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Reader Safety Glasses
Frame Color Clear
Magnification Power +1.75
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400
Frame Type Rimless
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Cancer and Reproductive Harm