Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Zeocin^t, 100 mg/ml in HEPES, sterile-filtered

Catalog No. AAJ67140XF
Change view
Click to view available options
Quantity:
1 mL
5 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67140XF 1 mL
AAJ671408EQ 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67140XF Supplier Thermo Scientific Chemicals Supplier No. J67140XF
Add to Cart

Zeocin (phleomycin, CAS # 11031-11-1) is a broad-spectrum antibiotic that acts by intercalating DNA.

Zeocin, a popular and effective antibiotic for the selection of vectors bearing the Sh ble gene in a variety of cells types (bacteria, eukaryotic microorganisms, plant and animal cells). Resistance to Zeocin is conferred by the Sh ble gene product, which inactivates Zeocin™ by binding to the antibiotic. Sh ble is a small gene, only 370bp in size, carried in a number of InvivoGen’s vectors. Zeocin is used as a selective agent in molecular genetics experiments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Zeocin (phleomycin) is a glycopeptide from the bleomycin family of antibiotics
  • Zeocin intercalates DNA, producing double-stranded breaks and preventing cell division
  • It can chelate some active metals including cobalt, iron, and copper
  • Zeocin has antineoplastic and antifungal properties

Application

  • Zeocin is useful for genetic selection experiments
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zeocin™
Packaging Plastic bottle
Quantity 1 mL
MDL Number MFCD28100499
Solubility Information Fully miscible.
Physical Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.