Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix 10X Plate Wash Buffer
SDP

Catalog No. 22157328 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-328 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-157-328 Supplier ZeptoMetrix Supplier No. 0801060
Add to Cart
Add to Cart

For washing microplates

  • Concentration: 10X

Specifications

Chemical Name or Material Microplate Wash Buffer
Concentration 10X
For Use With (Application) For washing microplates
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.