Learn More
Description
Includes
One 96 well microplate, 12mL detector antibody, 7mL of Rabbit IgG standard, 100mL assay diluent, 125mL of plate wash buffer, 12mL substrate, 12mL stop solution, 1 pack of microplate sealers, 1 plastic bag.
- Contains ready–to–use reagents
- Takes less than two hours to perform
- Microplate and detector antibody reacts with all subclasses of rabbit IgG
Specifications
Specifications
| Product Type | Immuno-Tek Rabbit IgG ELISA kit |
| Sufficient For | 96-well Microplate |
| For Use With (Application) | Designed for the measurement of Rabbit IgG in Rabbit Serum, Plasma or other biological fluids, the microplate and Detecto r Antibody in the kit have been specifically balanced to react uniformly with all subclasses of Rabbit IgG |
| Kit Contents | 1 96-well microplate, 1 pack of microplate sealers, 1 plastic bag, 100 mL assay diluent, 12 mL detector antibody, 12 mL stop solution, 12 mL substrate, 125 mL plate wash buffer, 7 mL of Rabbit IgG standard |
| Quantity | 96 Determinations |
| Storage Requirements | 2°C to 8°C |
Research purposes only. Not for in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
