Learn More
Description
Includes
One 96 well microplate, 12mL of Rat IgG detector antibody, 7mL of Rat IgG standard, 100mL assay diluent, 125mL of 10X plate wash buffer, 12mL substrate, 12mL stop solution, 1 pack of microplate sealers , 1 sealable plastic bag.
- Contains ready–to–use reagents
- Takes less than two hours to perform
- Microplate and detector antibody reacts with all subclasses of rat IgG
Specifications
Specifications
| Product Type | Immuno-Tek Rat IgG ELISA kit |
| Sufficient For | 96-well Microplate |
| For Use With (Application) | Designed for the measurement of rat IgG in rat serum, plasma, saliva, mucosa, cell culture fluid from rat cells or other biological sample, the assay contains ready-to -use reagents and takes less than two hours to perform, the microplate and detector antibody in the kit react with all subclasses of rat IgG |
| Quantity | 96 Determinations |
| Storage Requirements | 2°C to 8°C |
Research purposes only. Not for in vitro diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
