Learn More
ZeptoMetrix NATtrol™ Adenovirus Type 1
SDP
Catalog No. 22157761 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-157-761 1 mL
1 options
Catalog No. 22-157-761 Supplier ZeptoMetrix Supplier No. NATADV1ST
Qualitative molecular control
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Type Adenovirus Type 1 Control (qualitative)
For Use With (Application) Evaluate the performance of Nucleic Acid Tests for determination of the presence of Adenovirus Type 1 DNA and also for validation of clinical assays, development of diagnostic Tests and training of laboratory personnel
Quantity 1 mL