Learn More
Description
- Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Specifications
| Certifications/Compliance | ISO 13485 |
| Content And Storage | 2°C to 8°C |
| Type | Adenovirus Type 7A Control (qualitative) |
| For Use With (Application) | Evaluate the performance of Nucleic Acid Tests for determination of the presence of Adenovirus Type 7A DNA and also for validation of clinical assays, development of diagnostic Tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
