Learn More
Description
- Each control contains 6 x 0.5mL vials of NATtrol™ Influenza A/B or Influenza A H1N1 (2009) or Coxsackie virus A9
- Supplied in purified protein matrix identical to the composition of true clinical specimen
- Designed to evaluate the performance of nucleic acid tests for determination of presence of respiratory virus nucleic acids
Contain intact organisms and should be run in a manner identical to that used for clinical specimens.
Quality control of clinical assays and training of laboratory personnel
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Influenza A/NewCaledonia/20/99 Nucleic Acid, Influenza B/Florida/02/07 Nucleic Acid |
| Type | Influenza External Run Control |
| For Use With (Application) | Quality control of clinical assays and training of laboratory personnel |
| Quantity | 6 x 0.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

