Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix RETROtek SIV p27 Antigen ELISA Kit
SDP

Catalog No. 22156775 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
5 x 96 Determinations
96 Determinations
Sufficient For:
5 x 96 Microplate Wells
96-well Microplate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
22-156-775 96 Determinations 96-well Microplate
22-157-366 5 x 96 Determinations 5 x 96 Microplate Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-156-775 Supplier ZeptoMetrix Supplier No. 0801169
Add to Cart
Add to Cart

Includes

SIV p27 Antibody Coated Microplate for 96 determinations,1 plate: 12 x 8 well strips
SIV p27 Detector Antibody, 1 vial, lyophilized: Contains biotin-labelled polyclonal antibody to SIV, milk, Tween™, normal human serum and PBS
Detector Antibody Diluent: 25mL
SIV p27 Antigen Standard, 0.5mL: Contains detergent-disrupted, heat-inactivated viral antigen, goat serum, TritonX-100™ and sodium azide
Lysing Buffer, 5mL: Triton X-100™ in PBS and 2-chloroacetamide
Streptavidin peroxidase, 0.3mL: Contains streptavidin conjugated to horseradish peroxidase, PBS, goat serum, and 2-chloroacetamide
Assay Diluent, 100mL: Contains goat serum, PBS, Triton X-100™ and 2-chloroacetamide
10X Plate Wash Buffer, 125mL: Contains PBS, Tween 20™ and 2-chloroacetamide
Substrate, 0.3mL: Contains tetramethylbenzidine (TMB) and dimethyl sulfoxide
Substrate Buffer, 50mL: Contains citrate/acetate buffer, hydrogen peroxide and 2-chloroacetamide.

Requires

Disposable gloves, Validated adjustable micropipettes, single and multichannel, Test tubes and racks for preparing specimen and control dilutions, Graduated cylinders and assorted beakers, Validated automatic microplate washer or manual vacuum aspiration equipment, Validated incubator for 37°C ±1°C, Validated microplate reader, Timer, 1% sodium hypochlorite as disinfectant. May be prepared from household bleach, Distilled or deionized water

Used to detect Simian Immunodeficiency Virus (SIV) p27 antigen in research specimens

  • Enzyme linked immunoassay used to monitor the purification and biochemical behavior of SIV

Specifications

Product Type RETROtek SIV p27 Antigen ELISA kit
Sufficient For 96-well Microplate
For Use With (Application) Used to detect Simian Immunodeficiency Virus (SIV) p27 Antigen in research specimens, including cell culture media, It can also be used to monito r the purification and biochemical behavior of SIV
Kit Contents Biotin-labelled Polyclonal Antibody to SIV, milk, Tween™, normal Human Serum, PBS, Detector antibody diluent, SIV p27 antibody coated microplate, 1 plate: 12 x 8-well strips, SIV p27 Detector Antibody, 1 vial, lyophilized
Quantity 96 Determinations
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.