Learn More
ZeptoMetrix SIV Western Blot Assay Kit
SDP
Catalog No. 22157381 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Product Type:
Simian Immunodeficiency Virus (SIV) Western Blot Kit
Quantity:
10 strip kit
10 strips
30 strip kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
22-157-381 Simian Immunodeficiency Virus (SIV) Western Blot Kit 30 strip kit
22-157-380 Simian Immunodeficiency Virus (SIV) Western Blot Kit 10 strip kit
22-157-382 Simian Immunodeficiency Virus (SIV) Western Blot Kit 10 strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-157-381 Supplier ZeptoMetrix Supplier No. 0801501
Add to Cart
Add to Cart
Includes
SIV Nitrocellulose Strips (10 or 30 strips), Non-Reactive Control (1 vial, 100μL), Reactive Control (1 vial, 100μL), Blotting Buffer (1 bottle, 125mL), 20X Wash Buffer (1 bottle, 100mL), Conjugate (1 vial, 100μL), Substrate (1 bottle, 75mL), Blotting Powder (1 packet, 10g) and Incubation Trays
Requires
Rocking platform, Aspirator with disinfectant trap, Disposable gloves, Calibrated adjustable micropipettes, Graduated cylinders and assorted beakers, Timer, 1% sodium hypochlorite as disinfectant. May be prepared from household bleach, Distilled or deionized water and Tweezers or forceps
For the in vitro detection of antibodies to SIV in serum or plasma
  • The assay utilizes nitrocellulose strips that contain SIVMAC viral proteins resulting from electrophoresis and electroblotting of detergent disrupted virus particles
  • A serum control band is included on each strip to minimize the risk of false negative results due to operational error
Specifications
Product Type Simian Immunodeficiency Virus (SIV) Western Blot Kit
Quantity 30 strip kit
Content And Storage 2°C to 8°C