Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521807 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521807 Supplier Invitrogen™ Supplier No. PA521807
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (82%), Pig (98%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZER1 probably acts as target recruitment subunit in the E3 ubiquitin ligase complex ZER1-CUL2-Elongin BC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZER1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene ZER1
Gene Accession No. Q7Z7L7, Q80ZJ6
Gene Alias C230075L19Rik; C9orf60; homolog of Zyg-11; Hzyg; Protein zer-1 homolog; RGD1310284; RP11-545E17.4; Zer1; zer-1 homolog; ZYG; ZYG homolog; zyg-11 homolog B-like protein; zyg-11 related cell cycle regulator; zyg-11 related, cell cycle regulator; ZYG11BL; Zyg11b-like protein
Gene Symbols ZER1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human ZER1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10444, 227693
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.