Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meccanica GM Divisione Medicale ZERO ZERO Prefilled Formalin Container, 60 mL

Catalog No. 22906014
Change view
Click to view available options
Color:
Blue
Gray
Green
Lilac
Orange
Pink
Translucent
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-906-014 Yellow
22-906-016 Green
22-906-015 Pink
22-906-013 Blue
22-906-011 Translucent
22-906-012 Orange
22-906-017 Gray
22-906-018 Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-906-014 Supplier Meccanica GM Divisione Medicale ZERO Supplier No. 6014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Meccanica GM Divisione Medicale ZERO Prefilled Formalin Container, 60 mL protects against exposure to harmful chemicals and vapors during routine fixation of specimens in formalin.

  • Reduces or eliminates exposure to formalin and harmful vapors
  • Comes in a variety of colored caps for easy specimen identification
  • Security seal
  • Dual chamber keeps formalin trapped until container is activated

Specifications

Product Type Pre-filled Specimen Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Capacity (Metric) 60 mL
Closure Material Plastic
Closure Type Threaded Cap
Color Yellow
Diameter (Metric) 4.445 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.