Learn More
Cayman Chemical C16 dIhydro CeramIde-d9d185mg
SDP
Supplier:  Cayman Chemical 393895 MG
Encompass_Preferred
A pleuromutilin antibiotic active against various bacteria including MRSA (MIC50s 0.006-1 Ug/ml) active against fluoroquinolone- and macrolide-resistant strains of N. gonorrhoeae (MIC90s 1 mg/L for all) reduces the number of CFUs in a mouse model of S. pneumoniae thigh infection from 10-40 mg/kg reduces LPS-induced production of TNF-a IL-6 IL-1B and GM-CSF BALF neutrophil infiltration and lung MMP-9 levels in a mouse model of LPS-induced neutrophilia
Catalog No. 50-291-8614
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.