Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Newtex™ Zetex™ 130 Series Heat Resistant Leather Palm Gloves - 14 in.
SDP

Catalog No. 1139254 Shop All Newtex Industries, Inc. Products
Change view
Click to view available options
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-392-54 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-54 Supplier Newtex™ Supplier No. 2100009

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from Zetex™ Fiberglass Fabric for protection from temperatures up to 600°F for extended periods of time. Leather on the palm and thumb increases abrasion resistance and dexterity.

Zetex TM gloves with leather palm deliver extra cut resistance

  • Premium wool liner
  • Double stitched and reinforced
  • Leather on the palm/thumb increases abrasion resistance and dexterity
  • Inorganic fibers will not produce hazardous outgassing

Specifications

Color White
Material Zetex Fiberglass Fabric
Size Universal
Temperature (English) Max. 600°F
Temperature (Metric) Max 315°C
For Use With (Application) Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments
Glove Cuff Style Open
Palm Type Leather
Series Zetex™ 130 Series
Length (English) 14 in.
Length (Metric) 35.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.