Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Newtex™ ZetexPlus™ 220 Series Double Palm Heat Resistant Mitts - 14 in.
SDP

Catalog No. 1139258 Shop All Newtex Industries, Inc. Products
Change view
Click to view available options
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-392-58 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-58 Supplier Newtex™ Supplier No. 2100040
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mitts are constructed with a double layer of ZetexPlus™ Vermiculite Fabric on the palm/thumb for greater durability and extended protection from temperatures up to 2000°F.

  • Vermiculite coating evenly dissipates heat across the surface of the mitt
  • Coating significantly increases temperature rating and abrasion resistance
  • Double layered palm/thumb increases heat protection and durability
  • Premium wool liner
  • Inorganic fibers will not produce hazardous outgassing

Specifications

Coating Material Vermiculite
Color Tan
Hand Ambidextrous
Material ZetexPlus Fiberglass Fabric
Size Universal
Temperature (English) Max. 2000°F
Temperature (Metric) Max 1095°C
For Use With (Application) Kiln/Furnace Operations, Heat Processing Facilities, and High Temperature Industrial Environments
Glove Cuff Style Gauntlet
Liner Material Wool
Series ZetexPlus™ 220
Length (English) 14 in.
Length (Metric) 35.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.