Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP42 Monoclonal Antibody (OTI12A7), TrueMAB™, OriGene
SDP

Catalog No. 501681816
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1816 Supplier OriGene Supplier No. TA807869
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rex1 is an exonuclease domain-containing transcription factor protein that can bind to Elongin. The Elongin complex stimulates the rate of transcription elongation by RNA polymerase II through suppression of transient pauses of the polymerase at various sites along the DNA template. Rex1 is a putative stem cell marker that is highly expressed in embryonic and adult stem cells.

Specifications

Antigen ZFP42
Applications Western Blot
Classification Monoclonal
Clone OTI12A7
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZFP42
Gene Accession No. Q96MM3
Gene Alias REX-1, REX1, ZNF754, zfp-42
Gene Symbols ZFP42
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-235 of human ZFP42 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132625
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.