Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFP57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598857
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598857 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598857 Supplier Invitrogen™ Supplier No. PA598857
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZFP57 Polyclonal Antibody for ELISA, Western Blot

The protein encoded by this gene is a zinc finger protein containing a KRAB domain. Studies in mouse suggest that this protein may function as a transcriptional repressor. Mutations in this gene have been associated with transient neonatal diabetes mellitus type 1 (TNDM1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFP57
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZFP57
Gene Accession No. Q8C6P8, Q9NU63
Gene Alias bA145L22; bA145L22.2; C6orf40; DADB-25P22.4; G19; TNDM1; ZFP57; Zfp-57; ZFP57 zinc finger protein; zinc finger protein 57; Zinc finger protein 57 homolog; zinc finger protein 698; ZNF698
Gene Symbols ZFP57
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 57 homolog protein (183-295AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22715, 346171
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.