Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZFYVE9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597180 Supplier Invitrogen™ Supplier No. PA597180
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MENYFQAEAY NLDKVLDEFE QNEDETVSST LLDTKWNKIL DPPSHRLSFN PTLASVNESA VSNESQPQLK VFSLAHSAPL TTEEEDHCAN GQDCNLNPEI ATMWIDENAV AEDQLIKRNY SWDDQCSAVE VGEKKCGNLA CLPDEKNVLV VAVMHNCDKR TLQNDLQDCN NYNSQSLMDA FSCSLDNENR QTDQFSFSIN ESTEKDMNSE KQMDPLNRPK TEGRSVNHLC PTSSDSLASV CSPSQLKDDG SIGRDPSMSA; Positive Samples: LO2, HeLa; Cellular Location: Cytoplasm, Early endosome membrane.

Early endosomal protein that functions to recruit SMAD2/SMAD3 to intracellular membranes and to the TGF-beta receptor. Plays a significant role in TGF-mediated signaling by regulating the subcellular location of SMAD2 and SMAD3 and modulating the transcriptional activity of the SMAD3/SMAD4 complex. Possibly associated with TGF-beta receptor internalization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Zfyve9
Gene Accession No. O95405
Gene Alias E030027L17; hSARA; MAD, mothers against decapentaplegic homolog interacting protein, receptor activation anchor; MADH-interacting protein; MADHIP; Mothers against decapentaplegic homolog-interacting protein; Novel serine protease; NSP; PPP1R173; protein phosphatase 1, regulatory subunit 173; Receptor activation anchor; SARA; Smad anchor for receptor activation; SMADIP; ZFYVE9; Zinc finger FYVE domain-containing protein 9; zinc finger FYVE-type containing 9; zinc finger, FYVE domain containing 9
Gene Symbols Zfyve9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ZFYVE9 (NP_0047902).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9372
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.