Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZG16B Recombinant Human Monoclonal Antibody (6E5)
GREENER_CHOICE

Catalog No. FEMA550079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550079 Supplier Invitrogen™ Supplier No. MA550079
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

ZG16B Recombinant Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZG16B
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone 6E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ZG16B
Gene Accession No. Q96DA0
Gene Alias EECP; endocrine and exocrine protein; HRPE773; jacalin-like lectin domain containing 2; JCLN2; pancreatic adenocarcinoma upregulated factor; PAUF; PRO1567; UNQ773/PRO1567; ZG16B; Zymogen granule protein 16 homolog B; zymogen granule protein 16B
Gene Symbols ZG16B
Host Species Human
Immunogen Recombinant Human ZG16B protein.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124220
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.