Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIC1/ZIC2/ZIC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536307 Supplier Invitrogen™ Supplier No. PA536307
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 51 kDa. Purity is >95% by SDS-PAGE.

Zic1 encodes a zinc finger protein expressed in the developing or matured central nervous system in a highly restricted manner. Zic is expressed in granule cells that make synaptic contact with Purkinje cells. Zic1 is a gene critical to cerebellar pattern formation. The expression of Zic genes is first detected at gastrulation and at neurulation, becoming restricted to the dorsal neural ectoderm and the dorsal paraxial mesoderm. The Zic1 gene has been mapped to chromosome 9 in mouse. The 5' flanking region of the Zic1 gene contains a region-specific enhancer determined to be essential in in-vivo and in-vitro deletion analysis. The temporal profile of mRNA expression differs for each of the Zic gene products. The Drosophila odd-paired gene is highly homologous to the Zic gene family. Zic2 and Zic3 are highly similar genes. Zic2 is essential for the formation of the brain and Zic-3 is important for right and left axis formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC1/ZIC2/ZIC3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ZIC3
Gene Accession No. O60481, O95409, P46684, Q15915, Q62520, Q62521
Gene Alias Bn; CRS6; GENA 29; heterotaxy 1; HPE5; HTX; HTX1; Ka; Ku; kumba; odd-paired homolog; RGD1561261; VACTERLX; Zic; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); zic protein member 1; Zic1; Zic2; Zic3; Zinc finger protein 201; zinc finger protein 203; Zinc finger protein of the cerebellum 1; zinc finger protein of the cerebellum 2; Zinc finger protein of the cerebellum 3; zinc finger protein ZIC 1; Zinc finger protein ZIC 2; zinc finger protein ZIC 3; ZNF201; ZNF203
Gene Symbols ZIC1, ZIC2, ZIC3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 318-366 of human Zic1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22771, 22772, 22773, 361096, 367944, 64618, 7545, 7546, 7547
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.