Learn More
Invitrogen™ ZIC2 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 88% homologous in Xenopus and Chicken.
Specifications
Specifications
| Antigen | ZIC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | ZIC2 |
| Gene Accession No. | O95409, Q62520 |
| Gene Alias | cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 2; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; zinc finger protein ZIC 2; zinc finger protein Zic2 |
| Gene Symbols | ZIC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.