Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581131 Supplier Thermo Scientific Supplier No. PA581131
Only null left
Add to Cart
Add to Cart

ZIC3 Polyclonal Antibody

ZIC3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Zic3
Gene Accession No. O60481
Gene Alias Bn; heterotaxy 1; HTX; HTX1; Ka; RGD1561261; VACTERLX; ZIC; Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zic3; Zinc finger protein 203; zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201; ZNF203
Gene Symbols Zic3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human ZIC3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7547
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.