Learn More
TARGETMOL CHEMICALS INC Aticaprant 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ008210MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aticaprant (CERC-501) is a potent and centrally-penetrant antagonist of the kappa-opioid receptor (Ki 0.807 nM).[2] Purity 99.93%
Catalog No. 50-288-0423
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.