Learn More
Creative Biomart Recombinant Canine H3N2 Antigen, His-tagged
Supplier:  Creative Biomart ZIK441
Encompass_Preferred
Canine H3N2 produced in Sf9 Baculovirus cells is a single, glycosylated polypeptide chain containing 336 amino acids (18-344a.a.) and having a molecular mass of 36.9kDa. (Molecular size on SDS-PAGE wi
Catalog No. 50-208-4819
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.