Learn More
Bioss KD-Validated CD146 Rabbit Reco
SDP
Supplier:  Bioss BSM90119R
Encompass
Melanoma Cell Adhesion Molecule Primary Antibody Unconjugated Public Immunogen Range 600-646/646 Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB FCM IC Concentration Lot dependent Size 50 ul Storage Buffer Supplied in PBS (pH 7 4) containing 50% glycerol and 0 02% sodium azide
Catalog No. 50-000-64908
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.