Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus Envelope (strain Zika SPH2015) Recombinant Rabbit Monoclonal Antibody (033), HRP
GREENER_CHOICE

Catalog No. MA560379
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA560379 100 μg
MA560380 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA560379 Supplier Invitrogen™ Supplier No. MA560379
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Zika Virus Envelope (strain Zika SPH2015) Recombinant Monoclonal Antibody for ELISA

As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. and Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus Envelope (strain Zika SPH2015)
Applications ELISA
Classification Recombinant Monoclonal
Clone 033
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene Alias Zika Envelope
Host Species Rabbit
Immunogen Recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/Envelope protein (Domain III) protein (Val593-Lys699).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.