Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zika Virus NS2B (strain H/PF/2013) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143439 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143439 Supplier Invitrogen™ Supplier No. PA5143439
  • $493.00
    $457.65 / Each of 1

    Save $35.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: Zika virus-infected Vero, Zika virus NS2B-transfected 293T.

Zika virus (ZIKV) outbreaks are linked to severe neurological disorders ZIKV relies on its NS2B/NS3 protease for polyprotein processing; hence, this enzyme is an attractive drug target NS2B/NS3 viral protease, which, together with host cell proteases, cleaves the viral polyprotein into the individual proteins required for viral replication [DOI: 10.1126/science aag2419].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zika Virus NS2B (strain H/PF/2013)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias Zika virus Non-structural protein 2B; Zika virus NS2B; ZIKV Non-structural protein 2B; ZIKV NS2B
Host Species Rabbit
Immunogen Full length Zika virus NS2B recombinant protein. (Zika virus (strain H/PF/2013)).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.