Learn More
Cayman Chemical SMPD1humnrecombInt 50ug
SDP
Supplier:  Cayman Chemical 4108050 UG
Encompass_Preferred
Source Recombinant human C-terminal His-tagged PSGL-1 expressed in HEK293 cells Amino acids 18-295 MW 30.5 kDa
Catalog No. 50-294-4727
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.