Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zika Virus (SPH2015) Envelope anti-Virus, HRP, Antibody Pair, Novus Biologicals™
SDP

Catalog No. NB183334 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183334 5 Plates
NB183333 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB183334 Supplier Novus Biologicals Supplier No. NBP3067095PLATES
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antibody pair for Solid phase sandwich ELISA

Zika Virus (SPH2015) Envelope Monoclonal specifically detects Zika Virus (SPH2015) Envelope in Virus samples. It is validated for ELISA/Sandwich ELISA.

Specifications

Antigen Zika Virus (SPH2015) Envelope
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Conjugate HRP
Gene Alias Zika Envelope
Quantity 5 Plates
Regulatory Status RUO
Target Species Virus
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Type Antibody Pairs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.