Learn More
Zika Virus (SPH2015) NS1 Antibody (03), mFluor Violet 500 SE, R&D Systems™

Description
Specifications
Specifications
| Antigen | Zika Virus (SPH2015) NS1 |
| Applications | Sandwich ELISA Capture |
| Classification | Monoclonal |
| Clone | 03 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Sandwich ELISA Capture |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Zika NS1, ZIKV NS1, ZIKV-NS1 |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika Virus (Accession#: ALU33341.1; Val796-Leu1157). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.