Learn More
Selleck Chemical LLC Zileuton A-64077-20mg
SDP
Supplier:  Selleck Chemical LLC S144320MG
Encompass_Preferred
Zileuton (A-64077, Abbott 64077, ZYFLO) is an orally active inhibitor of 5-lipoxygenase, and thus inhibits leukotrienes (LTB4, LTC4, LTD4, and LTE4) formation, used to decrease the symptoms of asthma. Zileuton induces apoptosis while inhibits ferroptosis.
Catalog No. 50-239-0043
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.