Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Acetate Dihydrate, puriss. p.a., ACS Reagent, ≥99% (KT), Solstice

Catalog No. 60047784 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-784 1 kg Poly Bottle
60-047-785 250 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-784 Supplier Solstice Supplier No. 964591KG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Puriss. p.a., ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point 100°C
Color Gray
pH 6 to 8 (at 20°C)
Physical Form Crystalline
Quantity 1 kg
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3732513
Solubility Information In water 311g/L at 20°C
Formula Weight 219.51g/mol
Percent Purity ≥99% (KT)
Odor Vinegar like
Packaging Poly Bottle
Density 1.750g/cm3 at 20°C
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Hazard Statement P273-P305 + P351 + P338-P501
DOTInformation 9/PGIII
RTECSNumber 209-170-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.