Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Acetylacetonate, MP Biomedicals

Catalog No. icn219908 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219908 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219908 Supplier MP Biomedicals, Inc Supplier No. 0521990880

Chemical Identifiers

CAS 14024-63-6
Molecular Formula C10H14O4Zn
Molecular Weight (g/mol) 263.60
MDL Number MFCD00000035
InChI Key CYDXJXDAFPJUQE-FDGPNNRMSA-L
Synonym zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
PubChem CID 131675103
IUPAC Name zinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Quantity 100 g
Formula Weight 263.6
Chemical Name or Material Zinc Acetylacetonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.