Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Bromide, Anhydrous, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18607822 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-822 2.5 kg Amber Glass Bottle
18-607-824 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-822 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z10482.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Z1048, 7699-45-8, ZnBr2

Spectrum™ Chemical Zinc Bromide, Anhydrous, Reagent is a salt that has no color and share many similarities with zinc chloride. This inorganic compound is very soluble in water and is used as a Lewis acid and in an aqueous solution as a transparent shield against radiation in between two glass panes. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.19
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
IUPAC Name zinc(2+) dibromide
SMILES [Zn++].[Br-].[Br-]

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.